Learn More
BRAND™ BRANDplates™ Multiwell Plates for Cell Culture Inserts, - 6-well Specialized Plate
Shop All Brand Products
Click to view available options
Surface Treatment:
cellGrade plus
pureGrade S
Unit Size:
Pack of 10
Description
- BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
- Available with pureGrade™ S and cellGrade™ plus surfaces
- 6-well special plate design connects 4 wells into one large, elongated well, allowing simultaneous medium supply to all 4 inserts in a strip
- Ideal for insert strips with inlet channels, it also supports single inserts and combinations of 2 or 3 connected inserts
- Guide ridges for perfect insert fit
- Designated feeding ports for easy media changes, suitable for robotics/automation
- 10 plates per pack (individually wrapped with lid)
Surface Information:
- pureGrade™ S: Non-treated, hydrophobic surface with medium binding capacity (ELISA). Especially suited for bacteriological assays.
- cellGrade™ plus: Specifically designed for the cultivation of fastidious cell lines. Features carboxyl, hydroxy, and free amino groups on the surface, offering a protein-like composition that facilitates direct cell attachment and spreading.
Applications:
- Ideal for 3D tissue cultures
Compliance
ANSI/SLAS standards 1 and 4
Specifications
Specifications
| Closure Included | Yes |
| Bottom Shape | Flat |
| Capacity (Metric) | 10 mL |
| Color | Transparent |
| Culture Area | 855 mm2 |
| Material | Polystyrene |
| Sterility | Sterile |
| Surface Treatment | cellGrade plus |
| Type | Cell Culture Insert System |
| Well Count | 6 |
| Show More |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur