Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Description
- PTFE coating from point for 0.75 in.
- For dispensing both liquids and gases
- Precision-machined polymer plunger tip that creates leak-free seal
Available as replacement needle for installing into standard removable needle (RN) syringe; Needles sold separately
Specifications
Specifications
| Autoclavable | Autoclavable |
| Description | 1800 Series |
| Diameter (English) Inner | 0.057 in. |
| Diameter (English) Outer | 0.305 in. |
| Diameter (Metric) Inner | 1.46 mm |
| Diameter (Metric) Outer | 7.75 mm |
| Model | 1810 RN |
| Volume (Metric) | 100 μL |
| Connection Type | Removable |