Learn More
- Home
- Products
- Chromatography Syringes
- Manual Gas Tight Syringes
- Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Product Code. 10476323
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
Model:
1001
1002
1005
1010
1725
1750
1801 RN
1802
1805
1810 RN
1825
701
702
705
710
725
750
75RN
Unit Size:
Each
| Product Code. | Volume (Metric) | Model | unitSize |
|---|---|---|---|
| 10476323 | 250 μL | 725 | Each |
| 10335772 | 500 μL | 1750 | Each |
| 10759594 | 250 μL | 1725 | Each |
| 10079742 | 100 μL | 710 | Each |
| 11577864 | 500 μL | 750 | Each |
| 10374923 | 10 μL | 701 | Each |
| 11517624 | 25 μL | 702 | Each |
| 10438353 | 50 μL | 705 | Each |
| 10018132 | 10 mL | 1010 | Each |
| 10445112 | 5 mL | 1005 | Each |
| 11562515 | 250 μL | 1825 | Each |
| 10152702 | 1 mL | 1001 | Each |
| 10070892 | 2.5 mL | 1002 | Each |
| 10737453 | 50 μL | 1805 | Each |
| 10049492 | 25 μL | 1802 | Each |
| 10523765 | 100 μL | 1810 RN | Each |
| 10080234 | 10 μL | 1801 RN | Each |
| 10444805 | 5 μL | 75RN | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- ideal for dispensing volumes from 0.05 µL up to 5 µL
- The 7000 series is the only zero dead volume syringe
- The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Specifications
| Needle Gauge | 26s G |
| Model | 725 |
| Volume (Metric) | 250 μL |