Learn More
BRAND™ BRANDplates™ Multiwell Plates for Cell Culture Inserts, - 24-well Standard Plate
Product Code. 15503783 Shop All Brand Products
Change view
Click to view available options
Surface Treatment:
cellGrade plus
pureGrade S
Unit Size:
Pack of 10
Product Code. Surface Treatment unitSize
15503783 cellGrade plus Pack of 10
15620647 pureGrade S Pack of 10
2 options
Product Code. 15503783
Brand: BRAND™ 782890
Only null left
Add to basket
Add to basket
Sterile and individually wrapped and available with either untreated or cell culture treated surface.
  • BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
  • Available with pureGrade™ S and cellGrade™ plus surfaces
  • Includes 24 individually fillable wells that can be fitted with strips of 4 inserts and/or individual inserts
  • Guide ridges for perfect insert fit
  • Designated feeding ports for easy media changes, suitable for robotics/automation
  • 10 plates per pack (individually wrapped with lid)
Surface Information:
  • pureGrade™ S: Non-treated, hydrophobic surface with medium binding capacity (ELISA). Especially suited for bacteriological assays.
  • cellGrade™ plus: Specifically designed for the cultivation of fastidious cell lines. Features carboxyl, hydroxy, and free amino groups on the surface, offering a protein-like composition that facilitates direct cell attachment and spreading.
Applications:
  • Ideal for co-culture, secretions studies, and migrations studies
Compliance
ANSI/SLAS standards 1 and 4
TRUSTED_SUSTAINABILITY
Specifications
Closure Included Yes
Bottom Shape Flat
Capacity (Metric) 3.1 mL
Color Transparent
Culture Area 210 mm2
Material Polystyrene
Sterility Sterile
Surface Treatment cellGrade plus
Type Cell Culture Insert System
Well Count 24
For Use With (Application) Co-culture, secretion studies, migration studies
For Use With (Equipment) Insert system
Height (Metric) 19.5 mm