Learn More
BRAND™ BRANDplates™ Microplate Lids
Product Code. 15600499 Shop All Brand Products
Change view
Click to view available options
Plate Compatibility:
1536-well Standard Plate
384-well Plate
96-well Plate
96-well Standard Plate
No. per Pack:
50
100
Condensation Rings:
No
Yes
Unit Size:
Pack of 100
Pack of 50
Height (Metric):
4.5 mm
5.5 mm
8 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plate Compatibility No. per Pack Condensation Rings Unit Size Height (Metric)
15600499 96-well Plate 100 Yes Pack of 100 -
10337483 96-well Standard Plate 100 Yes Pack of 100 8 mm
10574584 96-well Standard Plate 100 No Pack of 100 8 mm
10468053 384-well Plate 50 No Pack of 50 4.5 mm
10574774 1536-well Standard Plate 50 No Pack of 50 5.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15600499 Supplier BRAND™ Supplier No. 782155
Estimated Shipment: 10-12-2026
CERTIFIED LIFE SCIENCE QUALITY plate covers for protection of samples, reaction batches, or cell cultures from contamination and evaporation.
  • Made from crystal-clear polystyrene
  • Cut-corner sets orientation of lid
  • Stackable
Lids with condensation rings available for 96-well standard plates
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Material Polystyrene
Sterility Non-sterile
No. of Wells 96
Condensation Rings Yes
No. per Pack 100
Evaporation Barrier No
For Use With (Application) For protection of samples, reaction batches or cell cultures from contamination and evaporation
Product Type Microplate Cover
Adhesive None
Temperature Range (Metric) <-20°C
Well Shape All well shapes
Plate Compatibility 96-well Plate
Cut-off Corners Yes