Learn More
BRAND™ BRANDplates™ Microplate Lids
Product Code. 10574774 Shop All Brand Products
Change view
Click to view available options
Plate Compatibility:
1536-well Standard Plate
384-well Plate
96-well Plate
96-well Standard Plate
No. per Pack:
50
100
Condensation Rings:
No
Yes
Height (Metric):
4.5 mm
5.5 mm
8 mm
Unit Size:
Pack of 100
Pack of 50
Product Code. Plate Compatibility No. per Pack Condensation Rings Height (Metric) unitSize
10574774 1536-well Standard Plate 50 No 5.5 mm Pack of 50
10337483 96-well Standard Plate 100 Yes 8 mm Pack of 100
10574584 96-well Standard Plate 100 No 8 mm Pack of 100
10468053 384-well Plate 50 No 4.5 mm Pack of 50
15600499 96-well Plate 100 Yes - Pack of 100
5 options
Product Code. 10574774
Brand: BRAND™ 782153
Only null left
Add to basket
Add to basket
CERTIFIED LIFE SCIENCE QUALITY plate covers for protection of samples, reaction batches, or cell cultures from contamination and evaporation.
  • Made from crystal-clear polystyrene
  • Cut-corner sets orientation of lid
  • Stackable
Lids with condensation rings available for 96-well standard plates
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Material Polystyrene
Sterility Non-sterile
No. of Wells 384/1536
Height (Metric) 5.5 mm
Condensation Rings No
No. per Pack 50
Evaporation Barrier No
Product Type Microplate Cover
Adhesive None
Temperature Range (Metric) <-20°C
Well Shape All well shapes
Plate Compatibility 1536-well Standard Plate
Cut-off Corners Yes