Learn More
Brady™ 4900 Series Thermal Transfer Printer Ribbons, Black
Description
- R4908, R4902, R4900, R4904 : core inside diameter is 1 in. (25.40mm), ink side out
- R4913, R4901 : core inside diameter is 0.5 in. (12.7mm)
Specifications
Specifications
| Product Type | Printer Ribbon |
| Color | Black |
| For Use With (Equipment) | Thermal Transfer Printer |
| Material | Polyester |
| Length (English) | 360 ft. |
| Width (English) | 4 in. |
| Length (Metric) | 110.00 m |
| Width (Metric) | 110 mm |
| Diameter (Metric) | 12.7 mm I.D. |
| Description | Ribbon Series: 4900 |
| Show More |