Learn More
3M™ 4432+ Lab Coats
Click to view available options
Size:
X-Large
2X-Large
Unit Size:
Pack of 50
Description
- Anti-static
- Front zip closing
- Low lint properties
- Reduced heat stress
- Knitted cuffs and collared
- Internal pocket on the wearer's left-hand side
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | X-Large |
| Gender | Unisex |
| Material | Polypropylene, Polyethylene Laminate |
| Color | Cool White |
| Cuff Style | Knit |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Seam Type | Serged |
| Certifications/Compliance | CE Category 1 |
| Show More |