Learn More
KGW Isotherm™ 4000mL Dewar Carrying Flask with Lid
Product Code. 10067740
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10067740 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10067740 Supplier KGW Isotherm™ Supplier No. 1224
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 09-12-2026
Specifications
Diameter (Metric) Outer 160mm
For Use With (Application) Cooling of small samples
Height (Metric) Inner 310mm
Height (Metric) 305mm
Includes Insulated lid, held by two spring clips, handle
Capacity (Metric) 4 L
Diameter (Metric) Inner 138 mm
Material Borosilicate Glass