Learn More
KGW Isotherm™ 4000mL Dewar Carrying Flask with Aluminum Jacket
Product Code. 10302621
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10302621 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10302621 Supplier KGW Isotherm™ Supplier No. 10223
Estimated Shipment: 04-01-2027
Spezifikation
Height (Metric) 375mm
Diameter (Metric) Outer 160mm
Capacity (Metric) 4 L
Diameter (Metric) Inner 138 mm
For Use With (Application) Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants
Height (Metric) Inner 310mm
Material Borosilicate Glass