Learn More
TubeWriter 400 - Standard Rack Top Print for 0.5 mL Microcentrifuge Tubes
Product Code. 30067435
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30067435 Labeling Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30067435 Supplier TubeWriter Supplier No. PT0500MCEL400
  • ONLINE EXCLUSIVE PRICE
    11735.00 NOK
    10131.71 NOK / Pack of 1
    Save 1603.29 NOK
    13% Off
Estimated Shipment: 12-01-2027
Standard rack for directly printing on the top of 0.5 mL microcentrifuge tubes
  • Standard printing rack designed for the 400
  • Prints and labels directly on the top of 0.5 mL microcentrifuge tubes
  • Rack ensures correct alignment and holds tubes securely
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80C and liquid nitrogen
  • Print withstands the harshest lab conditions
Specifications
Product Type Labeling Rack