Learn More
TubeWriter 400 - Standard Rack Side Print for 0.5-2 mL Cryogenic Vials and Screw Cap Tubes
Product Code. 30067407
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30067407 Labeling Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30067407 Supplier TubeWriter Supplier No. PS1800CVEL120400
  • ONLINE EXCLUSIVE PRICE
    11735.00 NOK
    10131.71 NOK / Pack of 1
    Save 1603.29 NOK
    13% Off
Estimated Shipment: 12-01-2027
Standard rack for directly printing on the side of 0.5 to 2 mL cryogenic vials and screw cap tubes
  • Standard printing rack designed for the 400
  • Prints and labels directly on the side of 0.5 to 2 mL cryogenic vials and screw cap tubes
  • Rack ensures correct alignment and holds vials securely
  • No time-consuming sticky labels or writing required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -807°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Product Type Labeling Rack