Learn More
Thermo Scientific™ 4-Way Flipper™ Racks
Product Code. 11530125 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
Unit Size:
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11530125 Fluorescent Pink Pack of 20
11520125 Fluorescent Orange Pack of 20
10321031 Fluorescent Green Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11530125 Supplier Thermo Scientific™ Supplier No. 8870
  • ONLINE EXCLUSIVE PRICE
    6410.00 NOK
    6200.35 NOK / Pack of 20
    Save 209.65 NOK
    3% Off
Estimated Shipment: 29-10-2026
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™4-Way Flipper™ Racks.
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™ 4-Way Flipper™ Racks.
TRUSTED_SUSTAINABILITY
Specifications
Description 4-Way Flipper Rack
Color Fluorescent Pink
Holds 0.5 mL, 15 mL, Tubes (50 mL)
Positions 32 x 0.5 mL, 32 x 1.5 mL, 12 x 15 mL or 4 x 50 mL tubes, each on a different side
Shape Rectangular
No. per Case 20 Racks
Product Type 4-way Flipper Rack