Learn More
4-Vinylbenzeneboronic acid, 97%
Product Code. p-7027165
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490128 1 g 1g
11400138 5 g 5g
11410138 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11490128 Supplier Thermo Scientific Alfa Aesar Supplier No. B23709.03
Request Bulk or Custom Format
CAS: 2156-04-9 | C8H9BO2 | 147.97 g/mol
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.
Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2156-04-9
Molecular Formula C8H9BO2
Molecular Weight (g/mol) 147.97
MDL Number MFCD00239441
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
PubChem CID 2734393
IUPAC Name (4-ethenylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C)C=C1
Specifications
Melting Point ∼190°C
Quantity 1 g
Beilstein 2829387
Solubility Information Soluble in ethanol,dimethylsulfoxide,tetrahydrofuran and diethyl ether. Slightly soluble in water.
Formula Weight 147.97
Percent Purity 97%
Chemical Name or Material 4-Vinylbenzeneboronic acid