Learn More
4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%
Product Code. p-7036720
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431188 1 g 1g
15441188 5 g 5g
15451188 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15431188 Supplier Thermo Scientific Alfa Aesar Supplier No. H55060.03
Request Bulk or Custom Format
CAS: 77123-57-0 | C12H14OSi | 202.33 g/mol
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
Notes
Store at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 77123-57-0
Molecular Formula C12H14OSi
Molecular Weight (g/mol) 202.33
MDL Number MFCD02093765
InChI Key UZQDUXAJFTWMDT-UHFFFAOYSA-N
Synonym 4-trimethylsilyl ethynyl benzaldehyde, 4-trimethylsilyl ethynylbenzaldehyde, 4-2-trimethylsilylethynyl benzaldehyde, 4-2-trimethylsilyl ethynyl benzaldehyde, benzaldehyde, 4-trimethylsilyl ethynyl, acmc-1bios, p-trimethylsilylethynylbenzaldehyde, 4-trimethylsilylethynylbenzaldehyde
PubChem CID 2771643
IUPAC Name 4-(2-trimethylsilylethynyl)benzaldehyde
SMILES C[Si](C)(C)C#CC1=CC=C(C=O)C=C1
Spezifikation
Melting Point 66°C to 70°C
Quantity 1 g
Formula Weight 202.33
Percent Purity 97%
Chemical Name or Material 4-[(Trimethylsilyl)ethynyl]benzaldehyde