Learn More
4-Trimethylsilyl-3-butyn-1-ol, 98%
Product Code. p-7035657
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15490128
Brand: Thermo Scientific Alfa Aesar H53376.06
Request Bulk or Custom Format
CAS: 2117-12-6 | C7H14OSi | 142.27 g/mol
4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2117-12-6
MDL Number MFCD00190217
Specifications
Boiling Point 97°C to 98°C (24mmHg)
Quantity 5 g
Molecular Formula C7H14OSi
Linear Formula HOCH2CH2C≡CSi(CH3)3
UN Number UN1993
Flash Point 60°C (140°F)
Synonym 4-trimethylsilyl-3-butyn-1-ol, 4-trimethylsilyl but-3-yn-1-ol, 3-butyn-1-ol, 4-trimethylsilyl, 4-trimethylsilyl-3-butyne-1-ol, 1-trimethylsilyl-1-butyn-4-ol, 3-butyn-1-ol,4-trimethylsilyl
InChI Key VSTMVAFCUIGHQI-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CCCO
IUPAC Name 4-trimethylsilylbut-3-yn-1-ol
Molecular Weight (g/mol) 142.27
PubChem CID 2760827
Formula Weight 142.27
Percent Purity 98%
Density 0.854
Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol