missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzene-1-carboximidamide hydrochloride dihydrate, 97%, Thermo Scientific™

Product Code. 10203432
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10203432 10 g 10g
10436872 1 g 1g
10596795 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10203432 Supplier Thermo Scientific Maybridge Supplier No. SPB01314EA
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-62-3
Molecular Formula C8H12ClF3N2O2
Molecular Weight (g/mol) 260.64
MDL Number MFCD00122370
InChI Key ZFGZRQQZWBYRRB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzamidine hydrochloride dihydrate, 4-trifluoromethyl benzimidamide hydrochloride dihydrate, 4-trifluoromethyl benzene-1-carboximidamide hydrochloride dihydrate, 4-trifluoromethyl benzenecarboximidamide dihydrate hydrochloride, benzenecarboximidamide,4-trifluoromethyl-, hydrochloride, hydrate 1:1:2, 4-trifluoromethyl benzamidine hydrochloridedihydrate, 4-trifluoromethylbenzamidine hydrochloride dihydrate, 4-trifluoromethyl benzamidine hydrochloride,dihydrate, 4-trifluoromethyl benzene-1-carboximidamide dihydrate hydrochloride, 4-trifluoromethyl benzene-1-carboximidamide-hydrogen chloride-water 1/1/2
PubChem CID 44630771
SMILES O.O.Cl.NC(=N)C1=CC=C(C=C1)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula HCl·H2O
Quantity 10 g
IUPAC Name 4-(trifluoromethyl)benzene-1-carboximidamide dihydrate hydrochloride
Formula Weight 242.63
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzene-1-carboximidamide hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.