Learn More
4-(Tri-n-butylstannyl)pyridine, 96%
Product Code. p-7033989
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15448087
Brand: Thermo Scientific Alfa Aesar H51435.03
Request Bulk or Custom Format
CAS: 124252-41-1 | C17H31NSn | 368.15 g/mol
4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.
Solubility
Not miscible in water. Soluble in ether, hexane and tetrahydrofuran.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 124252-41-1
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.15
MDL Number MFCD01319071
InChI Key UNEPXPMBVGDXGH-UHFFFAOYSA-N
PubChem CID 2737318
IUPAC Name tributyl(pyridin-4-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=NC=C1
Specifications
Quantity 1 g
UN Number UN2788
Synonym 4-tributylstannyl pyridine, pyridine,4-tributylstannyl, 4-tributylstannylpyridine, 4-tributylstannyl-pyridine, 4-tri-n-butylstannyl pyridine, pyridin-4-yl tributylstannane, pyridine, 4-tributylstannyl, 4-pyridyltributyltin, 4-pyridyl tributyltin, 4-tributyistannylpyridine
Solubility Information Not miscible in water. Soluble in ether,hexane and tetrahydrofuran.
Formula Weight 368.15
Percent Purity 96%
Chemical Name or Material 4-(Tri-n-butylstannyl)pyridine