missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenoxyacetyl chloride, 98%

Product Code. p-7024551
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11444007 10 g 10g
11454007 50 g 50g
2 options
This item is not returnable. View return policy

Product Code. 11444007

Brand: Thermo Scientific Alfa Aesar A19408.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90734-55-7 | C12H15ClO2 | 226.70 g/mol

4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alcohol, amine, bases, alkali metals, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90734-55-7
Molecular Formula C12H15ClO2
Molecular Weight (g/mol) 226.70
MDL Number MFCD00798568
InChI Key CFTNMBDQWVPSHI-UHFFFAOYSA-N
Synonym 4-tert-butylphenoxyacetyl chloride, 4-tert-butylphenoxy acetyl chloride, 2-4-tert-butylphenoxy acetyl chloride, 2-4-tert-butyl phenoxy acetyl chloride, pubchem14851, 4-tert-butylphenoxyacetyl cloride, para-t-butylphenoxyacetyl chloride, 4-t-butyl-phenoxy acetyl chloride, 4-tert-butyl-phenoxyacetyl chloride
PubChem CID 611195
IUPAC Name 2-(4-tert-butylphenoxy)acetyl chloride
SMILES CC(C)(C)C1=CC=C(OCC(Cl)=O)C=C1

Specifications

Boiling Point 130°C to 132°C (5 mmHg)
Refractive Index 1.5165
Quantity 10 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 226.71
Percent Purity 98%
Chemical Name or Material 4-tert-Butylphenoxyacetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.