Learn More
4-tert-Butylcyclohexanone, 99%
Product Code. 11467487
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11467487 500 g 500g
11457487 100 g 100g
2 options
Product Code. 11467487
Brand: Thermo Scientific Alfa Aesar A15612.36
Request Bulk or Custom Format
CAS: 98-53-3 | C10H18O | 154.253 g/mol
4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.
Solubility
Soluble in alcohol, ethanol (0.5g/10 mL). Insoluble in water.
Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-53-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00001642
InChI Key YKFKEYKJGVSEIX-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
PubChem CID 7392
IUPAC Name 4-tert-butylcyclohexan-1-one
SMILES CC(C)(C)C1CCC(=O)CC1
Specifications
Melting Point 47°C to 51°C
Density 0.893
Boiling Point 113°C to 116°C (20 mmHg)
Flash Point 96°C (204°F)
Quantity 500 g
Beilstein 507309
Solubility Information Soluble in alcohol,ethanol (0.5g/10 mL). Insoluble in water.
Formula Weight 154.25
Percent Purity 99%
Chemical Name or Material 4-tert-Butylcyclohexanone