Learn More
4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals
Product Code. 16607022 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 16607022
Brand: Thermo Scientific Acros 464350010
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 947725-04-4
Molecular Formula C12H17F3S
Molecular Weight (g/mol) 250.32
MDL Number MFCD11858620
InChI Key VRTQPEYVMHATOA-UHFFFAOYSA-N
IUPAC Name 5-tert-butyl-1,3-dimethyl-2-(trifluoro-λ⁴-sulfanyl)benzene
SMILES CC1=CC(=CC(C)=C1S(F)(F)F)C(C)(C)C
Specifications
Alpha Vector BUTYLDIMETHYLPHENYLSULFURTRIFLUORIDE
Melting Point 62°C to 68°C
Color Red to White
Boiling Point 92°C to 93°C
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
UN Number 2923
Sensitivity Moisture sensitive
Solubility Information insoluble aq. soln., soluble in hexane, acetonitrile, dichloromethane
Formula Weight 250.32
Physical Form Crystals or Powder
Chemical Name or Material 4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride