Learn More
4-Pyridylacetonitrile, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 13121-99-8 |
| Molecular Formula | C7H6N2 |
| MDL Number | MFCD03412601 |
| Quantity | 10 g |
| SMILES | N#CCC1=CC=NC=C1 |
| Molecular Weight (g/mol) | 118.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Pyridylacetonitrile |