Learn More
4-Pyridinecarboxylic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Pyridinecarboxylic Acid Ethyl Ester |
| CAS | 1570-45-2 |
| Molecular Formula | C8H9NO2 |
| Quantity | 100 g |
| Synonym | Isonicotinic Acid Ethyl Ester, 4-Carboethoxypyridine, 4-Ethoxycarbonylpyridine, Ethyl 4-Pyridinecarboxylate, Ethyl Isonicotinate, NSC 6854, gamma-Pyridinecarboxylic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C8H9NO2/c1-2-11-8(10)7-3-5-9-6-4-7/h3-6H,2H2,1H3 |
| SMILES | CCOC(=O)c1ccncc1 |
| IUPAC Name | ethyl pyridine-4-carboxylate |
| Molecular Weight (g/mol) | 151.16 |
| Formula Weight | 151.0633 |