Learn More
Labconco 4 place Microplate Rotor - CentriVap Pro
Click to view available options
Unit Size:
Each
Material (Coating):
PTFE
Description
- Compatible with CentriVap Pro Vacuum Concentrators
- Stainless steel material
- Optional PTFE coated
Specifications
Specifications
| Material | Stainless Steel |
| Type | Rotor |
| Rotor System | CentriVap Pro |
| Dimensions (L x W x H) | 25.4 x 15.75 x 7.62 cm (10 x 6.2 x 3 in.) |