Learn More
Gilac™ 4 Pivoting Wheel Dolly
Product Code. 17908511
Change view
Click to view available options
Includes:
Stainless Steel Wheel Housing
Steel Wheel Housing
Steel Wheel Housing, 2 Brakes
Color:
Black
Blue
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Color unitSize
17908511 Steel Wheel Housing Yellow Each
17908401 Steel Wheel Housing Black Each
17908791 Steel Wheel Housing Blue Each
17988391 Steel Wheel Housing, 2 Brakes Black Each
17968491 Stainless Steel Wheel Housing Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17908511 Supplier Gilac™ Supplier No. G170114
  • ONLINE EXCLUSIVE PRICE
    1265.25 NOK
    915.20 NOK / Pack of 1
    Save 350.05 NOK
    27% Off
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
4 Pivoting Wheel Dolly benefits from excellent stability and great manoeuvrability thanks to its 4 ball bearing mounted drive wheels.
  • Universal dolly on wheels.
  • Excellent stability and great manoeuvrability thanks to its 4 ball bearing mounted drive wheels.
  • 250 kg load capacity.
  • Outstanding quality, outclassing usual industry standards.
  • ABS injected base with solid frame guaranteeing unrivalled durability.
  • 100 mm diameter black PP wheels.
  • GILAC removable pulling bar available (as an option).
  • Compatible with all GILAC 600 x 400 and 400 x 300 mm containers and crates.
  • Dolly also available in black ABS-PC (G170144).
  • Stainless steel clevis recommended in humid environments (no damage to the bearings even in the presence of water).
Specifications
Type Dolly
Holds Containers and Crates
Load Bearing Capacity (English) 551 lb.
Load Bearing Capacity (Metric) 250 kg
Material ABS Plastic, Chrome-plated Steel
Caster Diameter (Metric) 100 mm
Color Yellow
Includes Steel Wheel Housing