Learn More
4-Phenyloxolan-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Phenyloxolan-2-one |
| CAS | 1008-73-7 |
| Molecular Formula | C10H10O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C10H10O2/c11-10-6-9(7-12-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2 |
| SMILES | C1C(COC1=O)C2=CC=CC=C2 |
| IUPAC Name | 4-phenyloxolan-2-one |
| Molecular Weight (g/mol) | 162.18 |
| Formula Weight | 162.0681 |