Learn More
4-(p-Tolyl)butyric acid, 97%
Product Code. 11467666
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11467666 25 g 25g
11447666 1 g 1g
11457666 5 g 5g
3 options
Product Code. 11467666
Brand: Thermo Scientific Alfa Aesar A14680.14
Request Bulk or Custom Format
CAS: 4521-22-6 | C11H14O2 | 178.231 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4521-22-6
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00021786
InChI Key IXWOVMRDYFFXGI-UHFFFAOYSA-N
Synonym 4-p-tolyl butanoic acid, 4-p-tolyl butyric acid, 4-4-methylphenyl butanoic acid, benzenebutanoic acid, 4-methyl, 4-p-tolylbutyric acid, 4-para-tolyl-butyric acid, 4-4-methylphenyl butyric acid, 4-p-tolyl butanoicacid, 4-p-tolylbutanoic acid, 4-p-tolyl-butyric acid
PubChem CID 78279
IUPAC Name 4-(4-methylphenyl)butanoic acid
SMILES CC1=CC=C(C=C1)CCCC(=O)O
Specifications
Melting Point 55°C to 59°C
Quantity 25 g
Beilstein 639705
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 4-(p-Tolyl)butyric acid