missing translation for 'onlineSavingsMsg'
Learn More

4-Octen-4-ylboronic acid pinacol ester, 98%, Thermo Scientific™

Product Code. p-7047743
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365535 250 mg 250mg
11355535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365535 Supplier Thermo Scientific Alfa Aesar Supplier No. L19699.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177949-95-0
Molecular Formula C14H27BO2
Molecular Weight (g/mol) 238.178
MDL Number MFCD03788750
InChI Key MVGKIRIFMYLBJA-UHFFFAOYSA-N
PubChem CID 71433658
IUPAC Name 4,4,5,5-tetramethyl-2-oct-4-en-4-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(=CCCC)CCC

Specifications

Refractive Index 1.448
Quantity 250 mg
Beilstein 9128681
Formula Weight 238.18
Percent Purity 98%
Chemical Name or Material 4-Octen-4-ylboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.