Learn More
4-Nitronaphthalen-1-ol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 605-62-9 |
| Molecular Formula | C10H7NO3 |
| MDL Number | MFCD02179392 |
| Quantity | 5 g |
| SMILES | OC1=C2C=CC=CC2=C([N+]([O-])=O)C=C1 |
| Molecular Weight (g/mol) | 189.17 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Nitronaphthalen-1-ol |