missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylbiphenyl, 98%, Thermo Scientific™

Product Code. 11458237
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458237 1 g 1g
11468237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458237 Supplier Thermo Scientific Alfa Aesar Supplier No. A17610.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10289-45-9
Molecular Formula C15H16
Molecular Weight (g/mol) 196.293
MDL Number MFCD00102114
InChI Key NAYIXKXYHOLMRC-UHFFFAOYSA-N
Synonym 4-propylbiphenyl, 4-propyl-1,1'-biphenyl, 4-n-propylbiphenyl, 4-propyl biphenyl, 4-propyl-1,1'-diphenyl, 1,1'-biphenyl, 4-propyl, 1-phenyl-4-propyl-benzene, 4-n-propyl biphenyl, pubchem2514, ksc492q3t
PubChem CID 2734126
IUPAC Name 1-phenyl-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 5°C to 17°C
Density 0.99
Boiling Point 109°C to 111°C (1.5 mmHg)
Refractive Index 1.5855
Quantity 1 g
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material 4-n-Propylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.