Learn More
4-(n-Propoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific™
Product Code. 11327539
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. Quantity unitSize
11327539 1 g 1g
11337539 10 g 10g
2 options
Product Code. 11327539
Brand: Thermo Scientific Alfa Aesar H27938.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 91062-38-3
Molecular Formula C10H13BO4
Molecular Weight (g/mol) 208.02
MDL Number MFCD06659852
InChI Key XJEONFAJOXSZDX-UHFFFAOYSA-N
Synonym 4-propoxycarbonyl phenylboronic acid, propyl 4-boronobenzoate, 4-n-propyloxycarbonylphenylboronic acid, n-propyl 4-boronobenzoate, 4-propoxycarbonylphenyl boronic acid, 4-propoxycarbonyl phenyl boronic acid, 4-n-propoxycarbonyl phenylboronic acid, 4-propoxycarbonyl benzene boronic acid, 4-n-propoxycarbonyl benzeneboronic acid
PubChem CID 23005364
IUPAC Name (4-propoxycarbonylphenyl)boronic acid
SMILES CCCOC(=O)C1=CC=C(C=C1)B(O)O
Specifications
Melting Point 121°C to 123°C
Quantity 1 g
Formula Weight 208.02
Percent Purity 97%
Chemical Name or Material 4-(n-Propoxycarbonyl)benzeneboronic acid