missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number 11416746
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
20651-71-2
C11H14O2
178.231
MFCD00002571
JFKUBRAOUZEZSL-UHFFFAOYSA-N
4-n-butylbenzoic acid, benzoic acid, 4-butyl, 4-butyl-benzoic acid, p-butylbenzoic acid, p-n-butylbenzoic acid, p-butyl benzoic acid, 4-butylbenzoicacid, p-n-butyl benzoic acid, acmc-209fcp, 4-n-butyl-benzoic acid
88631
4-butylbenzoic acid
CCCCC1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
20651-71-2
C11H14O2
178.231
MFCD00002571
JFKUBRAOUZEZSL-UHFFFAOYSA-N
4-n-butylbenzoic acid, benzoic acid, 4-butyl, 4-butyl-benzoic acid, p-butylbenzoic acid, p-n-butylbenzoic acid, p-butyl benzoic acid, 4-butylbenzoicacid, p-n-butyl benzoic acid, acmc-209fcp, 4-n-butyl-benzoic acid
88631
4-butylbenzoic acid
CCCCC1=CC=C(C=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C to 113°C
Quantity 5 g
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material 4-n-Butylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'