Learn More
4-(N-Boc-amino)-3-oxo-pentanoic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(N-Boc-amino)-3-oxo-pentanoic Acid Methyl Ester |
| CAS | 101669-78-7 |
| Molecular Formula | C11 H19 N O5 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C11H19NO5/c1-7(8(13)6-9(14)16-5)12-10(15)17-11(2,3)4/h7H,6H2,1-5H3,(H,12,15) |
| SMILES | COC(=O)CC(=O)C(C)NC(=O)OC(C)(C)C |
| IUPAC Name | methyl 4-[(2-methylpropan-2-yl)oxycarbonylamino]-3-oxopentanoate |
| Molecular Weight (g/mol) | 245.27 |
| Formula Weight | 245.1263 |