Learn More
4-Methylcyclohexanone, Sigma-Aldrich™
Specifications
Specifications
| CAS | 589-92-4 |
| Melting Point | -41°C |
| Density | 0.92 g/cm3 at 20°C |
| Boiling Point | 169°C to 171°C/1013 hPa |
| Molecular Formula | 4-(CH3)C6H9(=O) |
| MDL Number | MFCD00001643 |
| Quantity | 100 mL |
| Synonym | 4-Methylcyclohexanone, Tetrahydro-p-cresol, 4-Methylcyclohexanone |
| Molecular Weight (g/mol) | 112.17 |
| Grade | Research |