Learn More
4-Methylbenzyl Cyanide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methylbenzyl Cyanide |
| CAS | 2947-61-7 |
| Molecular Formula | C9H9N |
| Quantity | 2.5 g |
| Synonym | p-Tolylacetonitrile |
| InChI Formula | InChI=1S/C9H9N/c1-8-2-4-9(5-3-8)6-7-10/h2-5H,6H2,1H3 |
| SMILES | CC1=CC=C(CC#N)C=C1 |
| IUPAC Name | 2-(p-tolyl)acetonitrile |
| Molecular Weight (g/mol) | 131.17 |
| Formula Weight | 131.0735 |