Learn More
4-Methylamino Antipyrine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 4-Methylamino antipyrine |
| CAS | 519-98-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H15 N3 O |
| Quantity | 100 mg |
| Synonym | 1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3 H-pyrazol-3-one, Antipyrine, 4-(methylamino)- (6 CI,8 CI), 4-(Methylamino)antipyrine, 4-Methylaminophenazone, 4-Monomethylaminoantipyrine, 4-Monomethylaminophenazone, Methylaminoantipyrine, Methylaminophenazone, Monomethylaminoantipyrine, N-Methyl-4-aminoantipyrine, N-Methyl-4-aminophenazone, N-Methylaminoantipyrine, N-Methylaminophenazone, Noramidopyrine, Noraminopyrine, Metamizole Sodium Monohydrate Imp. C (EP) |
| InChI Formula | InChI=1 S/C12H15N3O/c1-9-11(13-2)12(16)15(14(9)3)10-7-5-4-6-8-10/h4-8,13 H,1-3H3 |
| SMILES | CNC1=C(C)N(C)N(C1=O)c2ccccc2 |
| IUPAC Name | 1,5-dimethyl-4-(methylamino)-2-phenylpyrazol-3-one |
| Molecular Weight (g/mol) | 217.27 |
| Show More |