Learn More
4-(Methylamino)-1-(3-pyridyl)-1-butanol-d3, TRC
Product Code. 30474982
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474982 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30474982 Supplier Toronto Research Chemicals Supplier No. TRCM286993100MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(Methylamino)-1-(3-pyridyl)-1-butanol-d3
CAS 1189642-32-7
Molecular Formula C10 2H3 H13 N2 O
Quantity 100 mg
InChI Formula InChI=1S/C10H16N2O/c1-11-6-3-5-10(13)9-4-2-7-12-8-9/h2,4,7-8,10-11,13H,3,5-6H2,1H3/i1D3
SMILES [2H]C([2H])([2H])NCCCC(O)c1cccnc1
IUPAC Name 1-pyridin-3-yl-4-(trideuteriomethylamino)butan-1-ol
Molecular Weight (g/mol) 183.27
Formula Weight 183.1451