Learn More
4-[Methyl(phenyl)sulfamoyl]benzeneboronic acid, 97%, Thermo Scientific™
Product Code. 15431596
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431596 1 g 1g
15441596 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15431596 Supplier Thermo Scientific Alfa Aesar Supplier No. H54789.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2096336-05-7
Molecular Formula C13H14BNO4S
Molecular Weight (g/mol) 291.13
MDL Number MFCD20265341
InChI Key PUWVRBKKJNKHLI-UHFFFAOYSA-N
Synonym 4-methyl phenyl sulfamoyl benzeneboronic acid
PubChem CID 73995963
Especificaciones
Quantity 1 g
Formula Weight 291.13
Percent Purity 97%
Chemical Name or Material 4-[Methyl(phenyl)sulfamoyl]benzeneboronic acid
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.