Learn More
4-Methyl-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide, TRC
Product Code. 30466314
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466314 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30466314 Supplier Toronto Research Chemicals Supplier No. TRCM142130500MG
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methyl-N-[4-methyl-3-[(4-(pyridin-3-yl)pyrimidin-2-yl)amino]phenyl]benzamide
CAS 152459-96-6
Molecular Formula C24 H21 N5 O
Quantity 500 mg
Synonym Benzamide, 4-methyl-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-, 4-Methyl-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, 4-Methyl-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide
InChI Formula InChI=1S/C24H21N5O/c1-16-5-8-18(9-6-16)23(30)27-20-10-7-17(2)22(14-20)29-24-26-13-11-21(28-24)19-4-3-12-25-15-19/h3-15H,1-2H3,(H,27,30)(H,26,28,29)
SMILES Cc1ccc(cc1)C(=O)Nc2ccc(C)c(Nc3nccc(n3)c4cccnc4)c2
IUPAC Name 4-methyl-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
Molecular Weight (g/mol) 395.46
Formula Weight 395.1746