missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-beta-styrylboronic acid pinacol ester, 98%

Product Code. 11335535
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335535 1 g 1g
11345535 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335535 Supplier Thermo Scientific Alfa Aesar Supplier No. L19698.03
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149777-84-4 | C15H21BO2 | 244.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149777-84-4
Molecular Formula C15H21BO2
Molecular Weight (g/mol) 244.141
MDL Number MFCD03788749
InChI Key HHBWKASJNTZJLB-ZHACJKMWSA-N
Synonym 4-methyl-beta-styrylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-4-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-4-methylphenyl ethenyl-1,3,2-dioxaborolane, 4-methyl-b-styrylboronic acid pinacol ester, 4-methyl-, a-styrylboronic acid pinacol ester, 2-e-4-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, beta-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl 4-methylstyrene
PubChem CID 11042960
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CC2=CC=C(C=C2)C

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Beilstein 7204616
Formula Weight 244.14
Percent Purity 98%
Chemical Name or Material 4-Methyl-beta-styrylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.