Learn More
4-Methyl-3,5-dinitrobenzeneboronic acid, 97%
Product Code. 11494340
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11494340
Brand: Thermo Scientific Alfa Aesar H25773.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 28249-49-2 | C7H7BN2O6 | 225.951 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28249-49-2
Molecular Formula C7H7BN2O6
Molecular Weight (g/mol) 225.951
MDL Number MFCD08060935
InChI Key DTWIWVTWNMEZOL-UHFFFAOYSA-N
Synonym 3,5-dinitro-4-methylbenzeneboronic acid, 4-methyl-3,5-dinitrophenyl boronic acid, 3,5-dinitro-4-methylphenylboronic acid, acmc-1cr7m, 3,5-dinitro-p-tolueneboronic acid, 3,5-dinitro-4-methylphenylboranic acid, 4-methyl-3,5-dinitrobenzeneboronic acid, p-tolueneboronic acid,3,5-dinitro-8ci, 3,5-dinitro-4-methylphenylboronic acid; 3,5-dinitro-p-tolueneboronic acid
PubChem CID 17998947
IUPAC Name (4-methyl-3,5-dinitrophenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)[N+](=O)[O-])C)[N+](=O)[O-])(O)O
Specifications
Melting Point ∼280°C (decomposition)
Quantity 1 g
Formula Weight 225.95
Percent Purity 97%
Chemical Name or Material 4-Methyl-3,5-dinitrobenzeneboronic acid