Learn More
4-Methyl-1-tetralone, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Methyl-1-tetralone is an important intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
4-Methyl-1-tetralone is an important intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Density | 1.078 |
Boiling Point | 95°C (1 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.56 |
Quantity | 1 g |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 160.22 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methyl-1-tetralone |