missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester, 98%, Thermo Scientific™

Catalog Number 15460596
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
334018-52-9
C14H18BClO4
296.55
MFCD06659928
SPVAFOQLYCMOAO-UHFFFAOYSA-N
methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-chloro-4-methoxycarbonylphenylboronic acid, pinacol ester, 3-chloro-4-methoxycarbonylphenylboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid, pinacol ester, methyl 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonyl-3-chlorophenylboronic acid, pinacol ester, methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzoate, methyl2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-3-chloro-4-methoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22493932
methyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
COC(=O)C1=C(Cl)C=C(C=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
334018-52-9
C14H18BClO4
296.55
MFCD06659928
SPVAFOQLYCMOAO-UHFFFAOYSA-N
methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-chloro-4-methoxycarbonylphenylboronic acid, pinacol ester, 3-chloro-4-methoxycarbonylphenylboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid, pinacol ester, methyl 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonyl-3-chlorophenylboronic acid, pinacol ester, methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzoate, methyl2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-3-chloro-4-methoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22493932
methyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
COC(=O)C1=C(Cl)C=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester is used as organic reagents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 48°C to 51°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 296.56
Percent Purity 98%
Chemical Name or Material 4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'