Learn More
4-Methoxy-3-nitrobenzoic acid, 98%, Thermo Scientific™
Product Code. 11424333
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424333 25 g 25g
11434333 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11424333 Supplier Thermo Scientific Alfa Aesar Supplier No. A10930.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-41-8
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007256
InChI Key ANXBDAFDZSXOPQ-UHFFFAOYSA-N
Synonym 4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid
PubChem CID 66640
SMILES COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]
Specifications
Melting Point 185°C to 190°C
Quantity 25 g
Beilstein 2115073
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid