Learn More
4-Methoxy-3-nitrobenzamide, 97%, Thermo Scientific™
Product Code. 11481377
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481377 25 g 25g
11461377 1 g 1g
11471377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11481377 Supplier Thermo Scientific Alfa Aesar Supplier No. B24905.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10397-58-7
Molecular Formula C8H8N2O4
Molecular Weight (g/mol) 196.162
MDL Number MFCD00017136
InChI Key PCQFJXUTKOUTRW-UHFFFAOYSA-N
PubChem CID 82603
IUPAC Name 4-methoxy-3-nitrobenzamide
SMILES COC1=C(C=C(C=C1)C(=O)N)[N+](=O)[O-]
Specifications
Melting Point 173°C to 176°C
Quantity 25 g
Synonym benzamide, 4-methoxy-3-nitro, benzamide,4-methoxy-3-nitro, 3-nitro-p-anisamide, p-anisamide, 3-nitro, acmc-20amfc, 4-methoxy-3-nitro-benzamide
Formula Weight 180.17
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-nitrobenzamide