missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitrophenol, 98%, Thermo Scientific™

Product Code. 11425433
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425433 50 g 50g
11415433 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425433 Supplier Thermo Scientific Alfa Aesar Supplier No. A13409.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1568-70-3
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.136
MDL Number MFCD00024247
InChI Key YBUGOACXDPDUIR-UHFFFAOYSA-N
Synonym phenol, 4-methoxy-2-nitro, 2-nitro-4-methoxyphenol, 4-hydroxy-3-nitroanisole, 4-methoxy-2-nitro-phenol, zlchem 182, pubchem16829, acmc-1c0rf, ksc494i1t, 4-methoxy-2-nitrophenol
PubChem CID 15285
IUPAC Name 4-methoxy-2-nitrophenol
SMILES COC1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 77°C to 80°C
Quantity 50 g
Solubility Information Soluble in methanol almost transparency.
Formula Weight 169.14
Percent Purity 98%
Chemical Name or Material 4-Methoxy-2-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.