Learn More
4-Methoxy-2-nitrophenol, 98%, Thermo Scientific™
Product Code. p-7021179
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415433 10 g 10g
11425433 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11415433 Supplier Thermo Scientific Alfa Aesar Supplier No. A13409.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol almost transparency.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1568-70-3
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.136
MDL Number MFCD00024247
InChI Key YBUGOACXDPDUIR-UHFFFAOYSA-N
Synonym phenol, 4-methoxy-2-nitro, 2-nitro-4-methoxyphenol, 4-hydroxy-3-nitroanisole, 4-methoxy-2-nitro-phenol, zlchem 182, pubchem16829, acmc-1c0rf, ksc494i1t, 4-methoxy-2-nitrophenol
PubChem CID 15285
IUPAC Name 4-methoxy-2-nitrophenol
SMILES COC1=CC(=C(C=C1)O)[N+](=O)[O-]
Specifications
Melting Point 77°C to 80°C
Quantity 10 g
Solubility Information Soluble in methanol almost transparency.
Formula Weight 169.14
Percent Purity 98%
Chemical Name or Material 4-Methoxy-2-nitrophenol