Learn More
4-Methoxy-2-methylaniline, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methoxy-2-methylaniline |
| CAS | 102-50-1 |
| Molecular Formula | C8H11NO |
| Quantity | 5 g |
| Synonym | 4-Methoxy-2-methylbenzenamine, 2-Amino-5-methoxytoluene, 2-Methyl-4-(methyloxy)aniline, 2-Methyl-4-anisidine, 2-Methyl-4-methoxyaniline, 2-Methyl-4-methoxybenzenamine, 2-Methyl-p-anisidine, 3-Methyl-4-aminophenol methyl ether, 4-Methoxy-o-toluidine, [2-Methyl-4-(methyloxy)phenyl]amine, m-Cresidine |
| InChI Formula | InChI=1S/C8H11NO/c1-6-5-7(10-2)3-4-8(6)9/h3-5H,9H2,1-2H3 |
| SMILES | COc1ccc(N)c(C)c1 |
| IUPAC Name | 4-methoxy-2-methylaniline |
| Molecular Weight (g/mol) | 137.18 |
| Formula Weight | 137.0841 |