Learn More
4-Methoxy-2,3,6-trimethylbenzaldehyde-d3, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methoxy-2,3,6-trimethylbenzaldehyde-d3 |
| CAS | 1216683-89-4 |
| Molecular Formula | C11 2H3 H11 O2 |
| Quantity | 25 mg |
| Synonym | 2,3,6-Trimethyl-4-(trideuteriomethoxy)benzaldehyde, 4-Methoxy-2,3,6-trimethylbenzaldehyde-d3, 2,3,6-Trimethyl-p-anisaldehyde-d3, 4-Methoxy-2,3,6-trimethylbenzaldehyde-D3 (methoxy-D3) |
| InChI Formula | InChI=1S/C11H14O2/c1-7-5-11(13-4)9(3)8(2)10(7)6-12/h5-6H,1-4H3/i4D3 |
| SMILES | [2H]C([2H])([2H])Oc1cc(C)c(C=O)c(C)c1C |
| IUPAC Name | 2,3,6-trimethyl-4-(trideuteriomethoxy)benzaldehyde |
| Molecular Weight (g/mol) | 181.25 |
| Formula Weight | 181.1182 |