Learn More
4-Methoxy-1-naphthoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methoxy-1-naphthoic Acid |
| CAS | 13041-62-8 |
| Molecular Formula | C12H10O3 |
| Quantity | 25 mg |
| Synonym | 4-Methoxy-1-naphthalenecarboxylic Acid, 4-Methoxy-1-naphthoic Acid, 4-Methoxy-alpha-naphthoic Acid |
| InChI Formula | InChI=1S/C12H10O3/c1-15-11-7-6-10(12(13)14)8-4-2-3-5-9(8)11/h2-7H,1H3,(H,13,14) |
| SMILES | COc1ccc(C(=O)O)c2ccccc12 |
| IUPAC Name | 4-methoxynaphthalene-1-carboxylic acid |
| Molecular Weight (g/mol) | 202.21 |
| Formula Weight | 202.063 |